missing translation for 'onlineSavingsMsg'
Learn More

IQUB Antibody, Novus Biologicals™

Product Code. 18439761 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
25 μL
Unit Size:
0.1mL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18439761 25 μL 25µL
18445112 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18439761 Supplier Novus Biologicals Supplier No. NBP23357325ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

IQUB Polyclonal antibody specifically detects IQUB in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen IQUB
Applications Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunocytochemistry/ Immunofluorescence 0.25-2 ug/mL
Formulation PBS (pH 7.2) and 40% Glycerol
Gene Accession No. Q8NA54
Gene Alias FLJ35834, IQ and ubiquitin-like domain-containing protein, IQ motif and ubiquitin domain containing, MGC149284, MGC149285
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to amino acids: MSNQQEKYEAQNIVNSTEESDDAFDTVTIPVPSEEPQESDQTEEHESGIEQFSESHAIHVEEQSDQSFSSLEPDNEQLMEEVISPRQVSYTPQHHEKQYAMQRPNDDSLAFLDKIKSVKESLQESVEDSLAT
Purification Method Immunogen affinity purified
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 154865
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.