missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IQUB Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA583972
This item is not returnable.
View return policy
Description
Immunogen sequence: MSNQQEKYEA QNIVNSTEES DDAFDTVTIP VPSEEPQESD QTEEHESGIE QFSESHAIHV EEQSDQSFSS LEPDNEQLME EVISPRQVSY TPQHHEKQYA MQRPNDDSLA FLDKIKSVKE SLQESVEDSL AT Highest antigen sequence indentity to the following orthologs: Mouse - 31%, Rat - 31%.
IQUB is a protein coding gene. May play roles in cilia formation and/or maintenance.
Specifications
| IQUB | |
| Polyclonal | |
| Unconjugated | |
| IQUB | |
| FLJ35834; IQ and ubiquitin-like domain-containing protein; IQ motif and ubiquitin domain containing; IQUB | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 154865 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry, Immunofluorescence | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8NA54 | |
| IQUB | |
| Recombinant protein corresponding to Human IQUB. Recombinant protein control fragment (Product #RP-92792). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction