Learn More
IRAK-1-4 Inhibitor I, MedChemExpress
Product Code. 16485648
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16485648
Brand: Medchem Express HY13329
MedChemExpress IRAK-1-4 Inhibitor I is an inhibitor of interleukin-1 receptor-associated kinase 1/4 (IRAK 1/4) with IC50s of 0.2 μM and 0.3 μM, respectively.
IRAK-1-4 Inhibitor I is an inhibitor of interleukin-1 receptor-associated kinase 1/4 (IRAK 1/4) with IC50s of 0.2 μM and 0.3 μM, respectively.
Specifications
Chemical Name or Material IRAK-1-4 Inhibitor I
CAS 509093-47-4
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C20H21N5O4
Quantity 10 mg
Synonym IRAK-1/4 Inhibitor I
Solubility Information DMF : 25 mg/mL (63.23 mM; ultrasonic and warming and heat to 60°C) ∣DMSO : 5 mg/mL (12.65 mM; ultrasonic and adjust pH to 3 with HCl) ∣Ethanol : < 1 mg/mL (insoluble) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C(C1=CC=CC([N+]([O-])=O)=C1)NC2=NC3=CC=CC=C3N2CCN4CCOCC4
Molecular Weight (g/mol) 395.41
Formula Weight 395.41
Percent Purity 99.2%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less
Research purposes only