Learn More
IRAK inhibitor 3, MedChemExpress
Product Code. 16411787
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Product Code. 16411787
Brand: Medchem Express HY13277
MedChemExpress IRAK inhibitor 3 is an interleukin-1 (IL-I) receptor-associated kinase (IRAK) kinase modulator extracted from patent WO2008030579 A2.
IRAK inhibitor 3 is an interleukin-1 (IL-I) receptor-associated kinase (IRAK) kinase modulator extracted from patent WO2008030579 A2.
Specifications
Chemical Name or Material IRAK inhibitor 3
CAS 1012343-93-9
Color Gray
Purity Grade Notes Research
Molecular Formula C21H21N5O4S
Quantity 50 mg
Solubility Information DMSO : 13 mg/mL (29.58 mM; Need ultrasonic and warming)
SMILES O=S(C1=CC=C(CNC2=NN3C(C4=CC(OC)=C(C=C4)OC)=CN=C3C=C2)C=C1)(N)=O
Molecular Weight (g/mol) 439.49
Formula Weight 439.49
Percent Purity 98.17%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less
Research purposes only