Learn More
IRAK inhibitor 3, MedChemExpress
MedChemExpress IRAK inhibitor 3 is an interleukin-1 (IL-I) receptor-associated kinase (IRAK) kinase modulator extracted from patent WO2008030579 A2.
€ 498.00 - € 1506.60
Specifications
Chemical Name or Material | IRAK inhibitor 3 |
---|---|
CAS | 1012343-93-9 |
Color | Gray |
Purity Grade Notes | Research |
Molecular Formula | C21H21N5O4S |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16478307
![]() |
Medchem Express
HY-13277 |
5 mg |
€ 498.00
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16425618
![]() |
Medchem Express
HY-13277 |
10 mg |
€ 600.75
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16411787
![]() |
Medchem Express
HY-13277 |
50 mg |
€ 1506.60
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IRAK inhibitor 3 is an interleukin-1 (IL-I) receptor-associated kinase (IRAK) kinase modulator extracted from patent WO2008030579 A2.Specifications
IRAK inhibitor 3 | |
Gray | |
C21H21N5O4S | |
O=S(C1=CC=C(CNC2=NN3C(C4=CC(OC)=C(C=C4)OC)=CN=C3C=C2)C=C1)(N)=O | |
439.49 | |
Research | |
COVID-19-immunoregulation |
1012343-93-9 | |
Research | |
DMSO : 13 mg/mL (29.58 mM; Need ultrasonic and warming) | |
439.49 | |
98.17% | |
Solid |
Safety and Handling
missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.