missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ IRAK1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-29523
This item is not returnable.
View return policy
Description
IRAK1 Polyclonal antibody specifically detects IRAK1 in Human samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| IRAK1 | |
| Polyclonal | |
| Tris-citrate/phosphate buffer, pH 7 to 8 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| EC 2.7.11, EC 2.7.11.1, interleukin-1 receptor-associated kinase 1, IRAK-1, IRAKPelle homolog, pelle | |
| Between 575 and 625 | |
| 100 μg | |
| Protein Kinase | |
| 3654 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction