missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IRAK1BP1 Antibody, mFluor Violet 500 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97199MFV500
This item is not returnable.
View return policy
Description
IRAK1BP1 Polyclonal antibody specifically detects IRAK1BP1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| IRAK1BP1 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin | |
| ActA binding protein 3,4921528N06Rik, AIP70, interleukin-1 receptor-associated kinase 1 binding protein 1, interleukin-1 receptor-associated kinase 1-binding protein 1, MGC138458, MGC138460, SIMPL | |
| Produced in rabbits immunized with E. coli-derived Human IRAK1BP1 fragment. | |
| 0.1 mL | |
| Protein Kinase | |
| 134728 | |
| Store at 4C in the dark. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction