missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IRAK1BP1 Antibody, Novus Biologicals™
Mouse Polyclonal Antibody
Brand: Novus Biologicals H00134728-B01P
This item is not returnable.
View return policy
Description
IRAK1BP1 Polyclonal antibody specifically detects IRAK1BP1 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| IRAK1BP1 | |
| Polyclonal | |
| Western Blot 1:500, Immunocytochemistry/ Immunofluorescence | |
| NP_001010844.1 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.4) | |
| ActA binding protein 3,4921528N06Rik, AIP70, interleukin-1 receptor-associated kinase 1 binding protein 1, interleukin-1 receptor-associated kinase 1-binding protein 1, MGC138458, MGC138460, SIMPL | |
| IRAK1BP1 (NP_001010844.1, 1 a.a. - 260 a.a.) full-length human protein. MSLQKTPPTRVFVELVPWADRSRENNLASGRETLPGLRHPLSSTQAQTATREVQVSGTSEVSAGPDRAQVVVRVSSTKEAAAEAKKSVCRRLDYITQSLQQQGVQAENITVTKDFRRVENAYHMEAEVCITFTEFGKMQNICNFLVEKLDSSVVISPPQFYHTPGSVENLRRQACLVAVENAWRKAQEVCNLVGQTLGKPLLIKEEETKEWEGQIDDHQSSRLSSSLTVQQKIKSATIHAASKVFITFEVKGKEKRKKHL | |
| 0.05 mg | |
| Protein Kinase | |
| 134728 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction