missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IRAK3 Antibody, Alexa Fluor™ 532, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76782AF532
This item is not returnable.
View return policy
Description
IRAK3 Polyclonal antibody specifically detects IRAK3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| IRAK3 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 532 | |
| ASRT5, EC 2.7.11.1, FLJ13601, IL-1 receptor-associated kinase M, interleukin-1 receptor-associated kinase 3, IRAK-3, IRAKM, IRAK-M | |
| Antibody was raised against a peptide corresponding to 16 amino acids near the carboxy terminus of human IRAK-M. The immunogen is located within the last 50 amino acids of IRAK-M. Amino Acid Squence: CSSKFSWDEYEQYKKE | |
| 0.1 mL | |
| Cytokine Research, Immunology, Innate Immunity, Protein Kinase | |
| 11213 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction