missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IRAK4 Antibody (413613) [Janelia Fluor™ 669], Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB39191JF669
This item is not returnable.
View return policy
Description
IRAK4 Monoclonal antibody specifically detects IRAK4 in Human samples. It is validated for Flow Cytometry (Intracellular Staining),Immunocytochemistry,CyTOF
Specifications
| IRAK4 | |
| Monoclonal | |
| Janelia Fluor 669 | |
| EC 2.7.11, EC 2.7.11.1, interleukin-1 receptor-associated kinase 4, IPD1, IRAK-4, NY-REN-64, REN64, Renal carcinoma antigen NY-REN-64 | |
| E. coli-derived recombinant human IRAK4, Met1-Ser460, Accession # Q9NWZ3 | |
| 0.1 mL | |
| Cytokine Research, Immunology, Innate Immunity, Phospho Specific, Protein Kinase | |
| 51135 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF | |
| 413613 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction