Learn More
IRAK4 Antibody (413613), mFluor Violet 500 SE, Novus Biologicals™
Rat Monoclonal Antibody
Brand:  Novus Biologicals FAB39191MFV500
Product Code. 18529757
  • € 595.00 / 0.10mL
Define Immunology Research on Your Terms With R&D Systems
Use promo code 24090
View all promotions
Description
IRAK4 Monoclonal antibody specifically detects IRAK4 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IRAK4
Monoclonal
mFluor Violet 500 SE
50mM Sodium Borate
Rat
0.1 mL
Cytokine Research, Immunology, Innate Immunity, Phospho Specific, Protein Kinase
51135
Store at 4C in the dark.
Flow Cytometry, Immunocytochemistry, CyTOF
413613
Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready
EC 2.7.11, EC 2.7.11.1, interleukin-1 receptor-associated kinase 4, IPD1, IRAK-4, NY-REN-64, REN64, Renal carcinoma antigen NY-REN-64
E. coli-derived recombinant human IRAK4, Met1-Ser460, Accession # Q9NWZ3