Learn More
IRAK4 Antibody (413613), PE, Novus Biologicals™
Rat Monoclonal Antibody
Brand:  Novus Biologicals FAB39191P
Product Code. 18604873
  • € 617.00 / 0.10mL
Description
IRAK4 Monoclonal antibody specifically detects IRAK4 in Human samples. It is validated for Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IRAK4
Monoclonal
PE
PBS
Rat
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
Human
Purified
Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF
413613
Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready
EC 2.7.11, EC 2.7.11.1, interleukin-1 receptor-associated kinase 4, IPD1, IRAK-4, NY-REN-64, REN64, Renal carcinoma antigen NY-REN-64
E. coli-derived recombinant human IRAK4, Met1-Ser460, Accession # Q9NWZ3
0.1 mL
Cytokine Research, Immunology, Innate Immunity, Protein Kinase
51135
Store at 4°C in the dark.