missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IRAK4 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA5144196

Product Code. 17999532

  • € 378.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

IRAK4 Polyclonal Antibody for Western Blot, ELISA, IP

IL-1 receptor associated kinases (IRAKs) are serine/threonine kinases involved in signaling via Toll/IL-1 pathways. Overexpression of IRAK-4 activates NF-kappa-B and MAPK pathways. IRAK-4 interacts with and phosphorylates IRAK-1. IRAK-4-deficient animals are completely resistant to the challenge with LPS. Animals and humans lacking IRAK-4 are impaired in their responses to viral and bacterial challenges. Members in IRAK/Pelle family play a central role in IL-1R/TLR mediated inflammatory responses and in innate immunity.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

IRAK4
Polyclonal
Unconjugated
IRAK4
8430405M07Rik; 9330209D03Rik; interleukin 1 receptor associated kinase 4; interleukin-1; interleukin-1 receptor-associated kinase 4; IPD1; Irak4; IRAK-4; NY-REN-64; REN64; renal carcinoma antigen NY-REN-64
Rabbit
Antigen affinity chromatography
RUO
51135
-20°C or -80°C if preferred
Liquid
ELISA, Immunoprecipitation, Western Blot
0.36 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9NWZ3
IRAK4
Recombinant Human Interleukin-1 receptor-associated kinase 4 protein (1-220AA).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.