missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IRE1 alpha Antibody, DyLight 650, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-2324C
This item is not returnable.
View return policy
Description
IRE1 alpha Polyclonal specifically detects IRE1 alpha in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.
Specifications
| IRE1 alpha | |
| Polyclonal | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, KnockDown Validated | |
| O75460 | |
| ERN1 | |
| This IRE1 alpha antibody was raised against a synthetic peptide within the human IRE1 alpha protein (within residues 700-800). [Swiss-Prot #O75460] | |
| Affinity Purified | |
| RUO | |
| Primary | |
| This antibody detects the levels of endogenous total IRE1 Alpha protein. | |
| Store at 4C in the dark. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown | |
| DyLight 650 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| EC 2.7.11, endoplasmic reticulum to nucleus signaling 1, Endoplasmic reticulum-to-nucleus signaling 1, ER to nucleus signalling 1, FLJ30999, hIRE1p, inositol-requiring 1, Inositol-requiring protein 1, IRE 1a, IRE1a, Ire1-alpha, IRE1MGC163279, IRE1P, MGC163277, protein kinase/endoribonuclease, serine/threonine-protein kinase/endoribonuclease IRE1 | |
| Rabbit | |
| 110 kDa | |
| 0.1 mL | |
| Apoptosis, Cell Biology, Diabetes Research, Hypoxia, Lipid and Metabolism, Neuroscience, Signal Transduction, Unfolded Protein Response | |
| 2081 | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto