missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IREB2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 23829-1-AP-150UL
This item is not returnable.
View return policy
Description
Iron regulatory protein 2 (IRP2) is a central post-transcriptional regulator of cellular and systemic iron metabolism. It is a cytosolic RNA-binding protein that binds to stem-loop structures. IRP2 is regulated by iron-mediated proteolysis. Hypoxia is important for normal tissue physiology as well as being a component of several pathophysiological conditions, including heart and cerebrovascular diseases and tumor growth. Because iron and oxygen are closely related, IRP2 also plays a role during hypoxia (low oxygen).Specifications
| IREB2 | |
| Polyclonal | |
| Unconjugated | |
| IREB2 | |
| ACO3, IRE BP 2, IREB2, Iron regulatory protein 2, IRP2, IRP2AD | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 3658, 64602 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P48200, Q811J3 | |
| IREB2 | |
| IREB2 Fusion Protein Ag20653 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction