missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IRF1 Antibody, DyLight 680, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-78763FR
This item is not returnable.
View return policy
Description
IRF1 Polyclonal antibody specifically detects IRF1 in Mouse samples. It is validated for Western Blot
Specifications
| IRF1 | |
| Polyclonal | |
| Western Blot | |
| interferon regulatory factor 1, MARIRF-1 | |
| The immunogen for this product maps to a region between residue 300 and 329 of mouse Interferon Regulatory Factor 1 using the numbering given in entry NP_032416.1 (GeneID 16362). | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 3659 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 680 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction