missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IRF1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-78762
This item is not returnable.
View return policy
Description
IRF1 Polyclonal specifically detects IRF1 in Mouse samples. It is validated for Immunoprecipitation.
Specifications
| IRF1 | |
| Polyclonal | |
| Unconjugated | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
| interferon regulatory factor 1, MARIRF-1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 3659 | |
| Store at 4C. Do not freeze. |
| Immunoprecipitation | |
| 1.0 mg/mL | |
| Immunoprecipitation 2-10 mcg/mg lysate | |
| NP_032416.1 | |
| IRF1 | |
| The immunogen for this product maps to a region between residue 250 and 300 of mouse Interferon Regulatory Factor 1 using the numbering given in entry NP_032416.1 (GeneID 16362). | |
| 100 μL | |
| Primary | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction