missing translation for 'onlineSavingsMsg'
Learn More

IRF1 Mouse anti-Human, PE, Clone: 20/IRF-1, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 566322

Product Code. 15824949

  • ONLINE EXCLUSIVE PRICE
    € 535.00
    € 533.18 / Each
    Save € 1.82
    0.34% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The 20/IRF-1 monoclonal antibody specifically binds to Interferon regulatory factor 1 (IRF1). IRF1 belongs to the human IRF family of transcription factors that contains nine members, IRF1-9. IRF1 is broadly expressed and its levels are upregulated in response to certain cytokines, interferons, bacterial or viral infections. IRF1 binds to specific DNA motifs such as IFN-stimulated response elements (ISRE) and regulates the activity of IFN-responsive genes. IRF1 plays roles in inflammation as well as innate and adaptive immune responses. It is involved in the regulation of cellular proliferation, Major Histocompatibility Complex (MHC) antigen expression, and the differentiation and effector functions of macrophages, dendritic cells, natural killer (NK) cells, and various T cell subsets. In addition, IRF1 may function as a tumor suppressor, since deletions in the IRF1 locus is associated with myelodysplasia and leukemias.

R-phycoerythrin (PE) is an accessory photosynthetic pigment found in red algae. It exists in vitro as a 240-kDa protein with 23 phycoerythrobilin chromophores per molecule. This makes PE one of the brightest fluorochromes for flow cytometry applications, but its photobleaching properties make it unsuitable for fluorescence microscopy.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

IRF1
20/IRF-1
PE
Interferon regulatory factor 1, IRF-1, MAR
Human IRF1 aa. 159-279
50 μg
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
0.2mg/mL
Aqueous buffered solution containing ≤0.09% sodium azide.
Mouse
Affinity Purified
RUO
Human
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.