missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IRF1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11335-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a DEAD box protein. DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division.Specifications
| IRF1 | |
| Polyclonal | |
| Unconjugated | |
| IRF1 | |
| interferon regulatory factor 1, IRF 1, IRF1, MAR | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 16362, 24508, 3659 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P10914, P15314, P23570 | |
| IRF1 | |
| IRF1 Fusion Protein Ag1879 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction