missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IRF2BP1 Monoclonal Antibody (OTI1H9)
GREENER_CHOICE

Product Code. 15907114
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15907114 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15907114 Supplier Invitrogen™ Supplier No. MA527520

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

IRF2BP1 Monoclonal Antibody for Western Blot, IHC (P)

IRF2BP1 acts as a transcriptional repressor. IRF2BP1 also acts as a transcriptional corepressor in a IRF2-dependent manner. This repression is not mediated at least in part by histone deacetylase activities.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IRF2BP1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI1H9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Irf2bp1
Gene Accession No. Q8IU81
Gene Alias 6330414O09Rik; C530033F24; DKFZP434M154; interferon regulatory factor 2 binding protein 1; interferon regulatory factor 2-binding protein 1; IRF-2-binding protein 1; IRF2BP1; IRF-2BP1; Probable E3 ubiquitin-protein ligase IRF2BP1; Probable RING-type E3 ubiquitin transferase IRF2BP1
Gene Symbols Irf2bp1
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 457-584 of IRF2BP1 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26145
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.