missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ IRF2BP2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-28956
Denne vare kan ikke returneres.
Se returpolicy
Beskrivelse
IRF2BP2 Polyclonal antibody specifically detects IRF2BP2 in Human samples. It is validated for Immunoprecipitation
Tekniske data
| IRF2BP2 | |
| Polyclonal | |
| Tris-citrate/phosphate buffer, pH 7 to 8 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 359948 | |
| Store at 4°C. Do not freeze. | |
| IgG |
| Immunoprecipitation | |
| Unconjugated | |
| interferon regulatory factor 2 binding protein 2, IRF-2-binding protein 2 | |
| between 225 and 275 | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion