missing translation for 'onlineSavingsMsg'
Learn More

IRF3 Antibody (3F10), DyLight 594, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP1-04308DL594

Product Code. 18492257

  • € 668.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

IRF3 Monoclonal antibody specifically detects IRF3 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/ Immunofluorescence, Gene Knock-Out, Single Cell Western
TRUSTED_SUSTAINABILITY
Specifications

Specifications

IRF3
Monoclonal
DyLight 594
interferon regulatory factor 3, IRF-3
Recombinant human IRF3 (108-166aa) purified from E. coli
0.1 mL
Immunology, Innate Immunity, Transcription Factors and Regulators
3661
Store at 4°C in the dark.
IgG1 κ
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, Gene Knock-Out
3F10
50mM Sodium Borate
Mouse
Protein G purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.