missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IRF3 Antibody (482205), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB4019H
This item is not returnable.
View return policy
Description
IRF3 Monoclonal antibody specifically detects IRF3 in Human samples. It is validated for Western Blot, Flow Cytometry (Intracellular Staining), CyTOF
Specifications
| IRF3 | |
| Monoclonal | |
| HRP | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry (Intracellular Staining), CyTOF | |
| 482205 | |
| Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| interferon regulatory factor 3, IRF-3 | |
| E. coli-derived recombinant human IRF3, aa 206-427, Accession # Q14653 | |
| 0.1 mL | |
| Immunology, Innate Immunity, Transcription Factors and Regulators | |
| 3661 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction