Learn More
IRF3 Antibody (482205), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB4019MFV500
Product Code. 18599799
  • € 663.00 / 0.10mL
Description
IRF3 Monoclonal antibody specifically detects IRF3 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IRF3
Monoclonal
mFluor Violet 500 SE
50mM Sodium Borate
Mouse
0.1 mL
Immunology, Innate Immunity, Phospho Specific, Transcription Factors and Regulators
3661
Store at 4C in the dark.
Western Blot, Flow Cytometry, CyTOF
482205
Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready
interferon regulatory factor 3, IRF-3
E. coli-derived recombinant human IRF3, aa 206-427, Accession # Q14653
RUO
Primary
Human
IgG2b