missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IRF3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 189.00
Specifications
| Antigen | IRF3 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1-4 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 2 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 2 μg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
IRF3 Polyclonal specifically detects IRF3 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| IRF3 | |
| Western Blot 1-4 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 2 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 2 μg/mL | |
| Polyclonal | |
| Rabbit | |
| Immunology, Innate Immunity, Transcription Factors and Regulators | |
| PBS with 0.02% Sodium Azide | |
| interferon regulatory factor 3, IRF-3 | |
| IRF3 | |
| IgG | |
| Affinity Purified | |
| At least three isoforms of IRF3 are known to exist. |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| NP_001562 | |
| 3661 | |
| Antibody was raised against a peptide corresponding to 14 amino acids near the carboxy terminus of human IRF3. The immunogen is located within amino acids 330 - 380 of IRF3. Amino Acid Squence: GESWPQDQPWTKRL | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| 47 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title