missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IRF3 Antibody, DyLight 650, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-78769C
This item is not returnable.
View return policy
Description
IRF3 Polyclonal antibody specifically detects IRF3 in Mouse samples. It is validated for Western Blot
Specifications
| IRF3 | |
| Polyclonal | |
| Western Blot | |
| interferon regulatory factor 3, IRF-3 | |
| The immunogen for this product maps to a region between residue 375 and 419 of mouse Interferon Regulatory Factor 3 using the numbering given in entry NP_058545.1 (GeneID 54131). | |
| 0.1 mL | |
| Immunology, Innate Immunity, Phospho Specific, Transcription Factors and Regulators | |
| 3661 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 650 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction