missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IRF3 Antibody (OTI2G3), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71042AF647
This item is not returnable.
View return policy
Description
IRF3 Monoclonal specifically detects IRF3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| IRF3 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| IRF3 | |
| Full-length protein expressed in 293T cell transfected with human IRF3 expression vector. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG2a |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI2G3 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| interferon regulatory factor 3, IRF-3 | |
| Mouse | |
| Affinity Purified | |
| Immunology, Innate Immunity, Phospho Specific, Transcription Factors and Regulators | |
| 3661.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction