missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IRF3 Antibody (PCRP-IRF3-1E6), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08854MFV500
This item is not returnable.
View return policy
Description
IRF3 Monoclonal antibody specifically detects IRF3 in Human samples. It is validated for Flow Cytometry
Specifications
| IRF3 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| PCRP-IRF3-1E6 | |
| Flow Cytometry | |
| interferon regulatory factor 3, IRF-3 | |
| Recombinant full-length human IRF3 protein (Uniprot: Q14653) | |
| 0.1 mL | |
| Immunology, Innate Immunity, Phospho Specific, Transcription Factors and Regulators | |
| 3661 | |
| Store at 4C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction