Learn More
IRF3 Mouse anti-Human, Clone: OTI5D2, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF500465
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
IRF-3 is encoded by the IRF-3 gene located on chromosome 19 in humans and belongs to the Interferon regulatory transcription factor (IRF) family. IRF-3 plays a critical role in the response of the immune system to viral infection. It binds to the Interferon stimulated response element (ISRE) of its downstream target genes such as Interferon alpha and beta to activate their transcription which eventually leads to immune modulatory actions of the type I Interferon (IFN) pathway. Following phosphorylation on Serine 396, IRF-3 has been shown to affiliate with CREB binding protein (CREBBP) and translocate to the nucleus which is critical for its ability to bind to DNA and act as a transcription factor. Additionally, phosphorylation associated nuclear translocation may also protect IRF-3 from degradation by the virus machinery.Specifications
| IRF3 | |
| Monoclonal | |
| Unconjugated | |
| IRF3 | |
| IIAE7 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 3661 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI5D2 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q14653 | |
| IRF3 | |
| Full-length protein expressed in 293T cell transfected with human IRF3 expression vector | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.