missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IRF3 Rabbit Recombinant Antibody, Proteintech
Rabbit Recombinant Antibody
Brand: Proteintech 80519-1-RR-100UL
This item is not returnable.
View return policy
Description
The IRF3 antibody from Proteintech is a rabbit recombinant antibody generated with peptide of human IRF3. This antibody recognizes human, mouse, zebrafish antigen. The IRF3 antibody has been validated for the following applications: WB, IHC, IF/ICC, ELISA analysis.Specifications
| IRF3 | |
| Recombinant Monoclonal | |
| 1000 μg/mL | |
| Rabbit | |
| Protein A purification | |
| RUO | |
| 3661 | |
| Store at -20°C. Stable for one year after shipment. | |
| Recombinant Antibody | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, ELISA | |
| 2H6 | |
| Unconjugated | |
| Peptide | |
| 100 μL | |
| Primary | |
| Human, Mouse, Zebrafish | |
| 50-60 kDa | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction