missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IRF4 Antibody (MUM1/8274R), CoraFluor™ 1, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-20718CL1
This item is not returnable.
View return policy
Description
IRF4 Monoclonal antibody specifically detects IRF4 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| IRF4 | |
| Monoclonal | |
| CoraFluor 1 | |
| interferon regulatory factor 4, IRF-4, LSIRFMUM1, Lymphocyte-specific interferon regulatory factor, Multiple myeloma oncogene 1, NF-EM5 | |
| Recombinant fragment (corresponding to C-terminal) of IRF4 protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Cancer | |
| 3662 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG κ |
| Immunohistochemistry (Paraffin) | |
| MUM1/8274R | |
| PBS | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction