missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IRF5 Antibody (10T1), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-04307B
This item is not returnable.
View return policy
Description
IRF5 Monoclonal specifically detects IRF5 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| IRF5 | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| IRF5 | |
| Recombinant human IRF5 (176-240 aa) purified from E. coli | |
| 0.1 mL | |
| Apoptosis, Death Receptor Signaling Pathway | |
| 3663 | |
| Human, Mouse | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 10T1 | |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| IBD14, interferon regulatory factor 5, IRF-5, SLEB10 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Anti-human IRF-5 mAb, clone 10T1, is derived from hybridization of mouse SP2/O myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human IRF-5 (176-240 aa). | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction