Learn More
IRF5 Antibody (903430) [DyLight 755], Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB8447Z
This item is not returnable.
View return policy
Description
IRF5 Monoclonal antibody specifically detects IRF5 in Mouse samples. It is validated for Flow Cytometry (Intracellular Staining), CyTOF
Specifications
| IRF5 | |
| Monoclonal | |
| DyLight 755 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry (Intracellular Staining), CyTOF | |
| 903430 | |
| Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| IBD14, interferon regulatory factor 5, IRF-5, SLEB10 | |
| E. coli-derived recombinant mouse IRF5, Ala208-Phe288, Accession # P56477 | |
| 0.1 mL | |
| Apoptosis, Death Receptor Signaling Pathway | |
| 3663 | |
| Store at 4°C in the dark. | |
| IgG2b |