missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IRF5 Antibody (903430) [Janelia Fluor™ 635], Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB8447JF635
This item is not returnable.
View return policy
Description
IRF5 Monoclonal antibody specifically detects IRF5 in Mouse samples. It is validated for Flow Cytometry (Intracellular Staining),CyTOF
Specifications
| IRF5 | |
| Monoclonal | |
| Janelia Fluor 635 | |
| IBD14, interferon regulatory factor 5, IRF-5, SLEB10 | |
| E. coli-derived recombinant mouse IRF5, Ala208-Phe288, Accession # P56477 | |
| RUO | |
| Primary | |
| Mouse | |
| Liquid |
| Flow Cytometry (Intracellular Staining), CyTOF | |
| 903430 | |
| 50mM Sodium Borate | |
| Rat | |
| 0.1 mL | |
| Apoptosis, Death Receptor Signaling Pathway | |
| 3663 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction