missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IRF5 Antibody (OTI1G7), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71043AF405
This item is not returnable.
View return policy
Description
IRF5 Monoclonal specifically detects IRF5 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| IRF5 | |
| Monoclonal | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| IRF5 | |
| Full length recombinant protein of human IRF5 (NP_116032) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI1G7 | |
| Western Blot, Immunohistochemistry | |
| IBD14, interferon regulatory factor 5, IRF-5, SLEB10 | |
| Mouse | |
| Affinity Purified | |
| Apoptosis, Death Receptor Signaling Pathway | |
| 3663.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction