missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IRF6 Antibody (OTI2C10), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71044APC
This item is not returnable.
View return policy
Description
IRF6 Monoclonal specifically detects IRF6 in Human, Mouse, Rat, Canine samples. It is validated for Flow Cytometry.
Specifications
| IRF6 | |
| Monoclonal | |
| APC | |
| PBS with 0.05% Sodium Azide | |
| IRF6 | |
| Full length recombinant protein of human IRF6 (NP_006138) produced in HEK293T cells. | |
| 0.1 mL | |
| 3664.0 | |
| Store at 4°C in the dark. |
| Flow Cytometry | |
| OTI2C10 | |
| Flow Cytometry | |
| interferon regulatory factor 6, IRF-6, LPS, OFC6PPS, PIT, Van der Woude syndrome, VWS, VWS1 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat, Canine | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction