missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IRF6 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17772374
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17772374 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17772374 Supplier Invitrogen™ Supplier No. PA5120979

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: 22Rv1, BT-474, HT-29, MCF7, 293T, Mouse liver, Mouse kidney, Rat liver. The target is usually found in the following locations: Cytoplasm, Nucleus. Immunogen sequence: DIPQPQGSII NPGSTGSAPW DEKDNDVDEE DEEDELDQSQ HHVPIQDTFP FLNINGSPMA PASVGNCSVG NCSPEAVWPK TEPLEMEVPQ APIQPFYSSP ELWISSLPMT DLDIKFQYRG KEYGQTMTVS NPQGCRLFYG DLGPMPDQEE LFGPVSLEQV KFPGPEHITN EKQKLFTSKL LDVMDR.

This gene encodes a member of the interferon regulatory transcription factor (IRF) family. Family members share a highly-conserved N-terminal helix-turn-helix DNA-binding domain and a less conserved C-terminal protein-binding domain. Mutations in this gene can cause van der Woude syndrome and popliteal pterygium syndrome. This protein is involved in palate formation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IRF6
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Irf6
Gene Accession No. O14896, P97431
Gene Alias AI876454; E230028I05Rik; interferon regulatory factor 6; IRF6; IRF-6; irf6 protein; IRF-6; transcription factor; LPS; mirf6; OFC6; PIT; PPS; PPS1; transcriptional factor; VWS; VWS1; zgc:63500
Gene Symbols Irf6
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 115-300 of human IRF6 (NP_006138.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 364081, 3664, 54139
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.