missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IRF7 Antibody (3D9), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-04309MFV610
This item is not returnable.
View return policy
Description
IRF7 Monoclonal antibody specifically detects IRF7 in Human samples. It is validated for Western Blot, ELISA
Specifications
| IRF7 | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| 3D9 | |
| Western Blot, ELISA | |
| interferon regulatory factor 7, interferon regulatory factor-7H, IRF-7, IRF7A, IRF-7H | |
| Recombinant human IRF7 (1-150aa) purified from E. coli (NP_001563). | |
| 0.1 mL | |
| Transcription Factors and Regulators | |
| 3665 | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction