missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ IRF7 Antibody (3D9), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-27934
This item is not returnable.
View return policy
Description
IRF7 Monoclonal antibody specifically detects IRF7 in Human samples. It is validated for ELISA,Immunohistochemistry,Western Blot
Specifications
| IRF7 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1 - 2 μg/mL, ELISA, Immunohistochemistry 0.5 - 1 μg/mL | |
| Q92985 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunohistochemistry, Western Blot | |
| 3D9 | |
| Unconjugated | |
| PBS (pH 7.4) | |
| interferon regulatory factor 7, interferon regulatory factor-7H, IRF-7, IRF7A, IRF-7H | |
| Recombinant human IRF7 (1-150aa) purified from E. coli. | |
| 0.05 mg | |
| Transcription Factors and Regulators | |
| 3665 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction