missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IRGM Antibody, DyLight 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-18254V
This item is not returnable.
View return policy
Description
IRGM Polyclonal antibody specifically detects IRGM in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence
Specifications
| IRGM | |
| Polyclonal | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| IFI1, immunity-related GTPase family M protein, immunity-related GTPase family, M, interferon-inducible protein 1, IRGM1, LPS-stimulated RAW 264.7 macrophage protein 47 homolog, LRG47, LRG-47, LRG-47-like protein, M1, MGC149263, MGC149264 | |
| Synthetic peptide from the N-terminal of Human IRGM | |
| 0.1 mL | |
| Autophagy | |
| 345611 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| DyLight 405 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction