missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Irisin/FNDC5 Antibody (936434) [PE/Atto594], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ FAB9420PEATT594
This item is not returnable.
View return policy
Description
Irisin/FNDC5 Monoclonal antibody specifically detects Irisin/FNDC5 in Human samples. It is validated for Flow Cytometry
Specifications
| Irisin/FNDC5 | |
| Monoclonal | |
| PE/ATTO 594 | |
| fibronectin type III domain containing 5, fibronectin type III domain-containing protein 5, FRCP2Fibronectin type III repeat-containing protein 2 | |
| Chinese hamster ovary cell line CHO-derived recombinant human Irisin/FNDC5, Asp32-Glu143, Accession # Q8NAU1 | |
| 0.1 mL | |
| Lipid and Metabolism, Neuroscience, Stem Cells | |
| 252995 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2b |
| Flow Cytometry | |
| 936434 | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction