missing translation for 'onlineSavingsMsg'
Learn More

Hach Lange™ Iron Cuvette Tests

Product Code. 10127309
Change view
Click to view available options
No. of Tests:
25
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Tests Unit Size
10127309 25 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10127309 Supplier Hach Lange™ Supplier No. LCK321
  • ONLINE EXCLUSIVE PRICE
    € 159.40
    € 146.48 / Pack of 1
    Save € 12.92
    8% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Order Info

dimProductType

TRUSTED_SUSTAINABILITY

Specifications

No. of Tests 25
Range 0.2 to 0.6mg/L Fe
Storage Requirements 2°C to 8°C
Test Method 1.10-Phenanthroline
Type Iron Cuvette Test
compliance-icons
Product Identifier
  • Eisen/Iron/Fer
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
Supplemental information
  • MIXTURE LIST-Contain: 1, 10-Phenanthrolinechloride monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.