missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ Iron Qualitative Test Paper
Shop All Macherey-Nagel Products
Click to view available options
Color Indicator:
Yellow/White to Red/Brown
Unit Size:
Pack of 100
Specifications
Specifications
| Color Indicator | Yellow/White to Red/Brown |
| Format | Strip |
| For Use With (Application) | Determination of Iron |
| Quantity | 100 Pack |
| Type | Qualitative Test Paper |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction