Learn More
Hach Lange™ Iron trace Cuvette Test
Product Code. 10273824
Change view
Click to view available options
Range:
0.01 to 1mg/L Fe
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Range Unit Size
10273824 0.01 to 1mg/L Fe Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10273824 Supplier Hach Lange™ Supplier No. LCK521
  • ONLINE EXCLUSIVE PRICE
    € 234.00
    € 160.48 / Pack of 1
    Save € 73.52
    31% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Order Info
dimRange
TRUSTED_SUSTAINABILITY
Specifications
No. of Tests 20
Range 0.01 to 1mg/L Fe
Storage Requirements 2°C to 8°C
Test Method 1.10-Phenanthroline
Type Iron Trace Cuvette Test
compliance-icons
Product Identifier
  • Iron, trace, cuvette test 50mm measuring range 0.
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P391-Collect spillage.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: 1,10-Phenanthroline,monohydrochloride,monohydrate, Ferricnitrate nonahydrate