missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Irx5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA568832
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 93%; Dog: 92%; Guinea Pig: 86%; Horse: 100%; Human: 93%; Mouse: 100%; Pig: 93%; Rabbit: 86%; Rat: 100%.
Establishes the cardiac repolarization gradient by its repressive actions on the KCND2 potassium-channel gene. Required for retinal cone bipolar cell differentiation. May regulate contrast adaptation in the retina and control specific aspects of visual function in circuits of the mammalian retina. Could be involved in the regulation of both the cell cycle and apoptosis in prostate cancer cells. Involved in craniofacial and gonadal development. Modulates the migration of progenitor cell populations in branchial arches and gonads by repressing CXCL12.
Specifications
| Irx5 | |
| Polyclonal | |
| Unconjugated | |
| IRX5 | |
| HMMS; Homeodomain protein IRX-2A; homeodomain protein IRXB2; iroquois homeobox 5; iroquois homeobox protein 5; Iroquois related homeobox 5 (Drosophila); iroquois-class homeodomain protein IRX-5; IRX2A; IRX-2a; IRX5; IRXB2; LOW QUALITY PROTEIN: iroquois-class homeodomain protein IRX-5 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 54352 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9JKQ4 | |
| IRX5 | |
| synthetic peptide directed towards the middle region of mouse Irx5. | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction