missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IRX6 Antibody (1A5), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00079190-M01
This item is not returnable.
View return policy
Description
IRX6 Monoclonal antibody specifically detects IRX6 in Human samples. It is validated for Western Blot, ELISA, Sandwich ELISA, KnockDown
Specifications
| IRX6 | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| iroquois homeobox 6, Iroquois homeobox protein 6Homeodomain protein IRXB3, iroquois homeobox protein 7, iroquois-class homeodomain protein IRX-6, IRX-3, IRX7IRXB3 | |
| IRX6 (NP_077311, 337 a.a. ∽ 446 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. PRLTMHYPCLEKPRIWSLAHTATASAVEGAPPARPRPRSPECRMIPGQPPASARRLSVPRDSACDESSCIPKAFGNPKFALQGLPLNCAPCPRRSEPVVQCQYPSGAEAG | |
| 0.1 mg | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Sandwich ELISA, KnockDown | |
| 1A5 | |
| Western Blot 1:500, ELISA, Sandwich ELISA, Knockdown Validated | |
| NP_077311 | |
| Mouse | |
| IgG purified | |
| RUO | |
| 79190 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction