Learn More
ISAB Solution NA, Reagecon™
Product Code. 11709165 Shop All Reagecon Products
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
Product Code. 11709165
Brand: Reagecon ISANA5
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Quantity 500 mL
compliance-icons compliance-icons
Product Identifier
  • SODIUM BASED ISA STANDARD
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : ammonium chloride, ammonia