Learn More
Invitrogen™ Iscalimab Recombinant Monoclonal Antibody
Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA559844
 View more versions of this product
Product Code. 30284450
  • € 1262.00 / 1mg
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Iscalimab (CFZ533) is a new, fully humanized monoclonal antibody that targets cluster of differentiation 40 (CD40), which can regulate the CD40-CD154 costimulatory pathway and prevent CD40 pathway signal transduction and CD40+ cell type activation. It is expected Become a new treatment for solid organs and related autoimmune diseases.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Iscalimab
Recombinant Monoclonal
Unconjugated
CFZ-533; NVP-CFZ533; OM11-62MF
Protein A
RUO
Human
Antibody
IgG1
ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
1 mg/mL
25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Human
1 mg
Primary
-20°C, Avoid Freeze/Thaw Cycles
Lyophilized