missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ISCU Monoclonal Antibody (OTI8H2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA803532S
This item is not returnable.
View return policy
Description
ISCU Monoclonal antibody specifically detects ISCU in Human samples. It is validated for Western BlotSpecifications
| ISCU | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P08118 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 4477 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI8H2 | |
| Unconjugated | |
| MSMB | |
| 2310020H20Rik; AA407971; HML; hnifU; iron-sulfur cluster assembly enzyme; iron-sulfur cluster assembly enzyme ISCU, mitochondrial; iron-sulfur cluster scaffold homolog; Iscu; IscU iron-sulfur cluster scaffold homolog; IscU iron-sulfur cluster scaffold homolog (E. coli); ISCU2; ISU2; Nifu; NifU-like N-terminal domain containing; nifU-like N-terminal domain-containing protein; NifU-like protein; NIFUN; RGD1309562 | |
| Purified prostate specific antigen (MSMB) from human seminal fluid | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction