missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ISCU Monoclonal Antibody (OTI8H2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA803532
This item is not returnable.
View return policy
Description
Iron-sulfur (Fe-S) clusters are necessary for several mitochondrial enzymes and other subcellular compartment proteins. They contain sulfur and iron, and are created via several steps that include cysteine desulfurases, iron donors, chaperones, and scaffold proteins. This gene encodes the two isomeric forms, ISCU1 and ISCU2, of the Fe-S cluster scaffold protein. Mutations in this gene have been found in patients with myopathy with severe exercise intolerance and myoglobinuria.Specifications
| ISCU | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9H1K1 | |
| ISCU | |
| Full length human recombit protein of human ISCU produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI8H2 | |
| Unconjugated | |
| ISCU | |
| 2310020H20Rik, HML, ISU2, NIFU, NIFUN, hnifU | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 23479 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction